Learn More
Keystone Cleanroom Products Autoclavable 1421B Tyvek™ Mini-Muffs
GREENER_CHOICE
Product Code. 30020313 Shop All Keystone Cleanroom Products Products
Click to view available options
Width (Metric):
1.3 to 2.5 cm
10.2 to 14 cm
14 to 20.3 cm
2.5 to 4.4 cm
20.3 to 30.5 cm
4.4 to 6.4 cm
6.4 to 10.2 cm
8.9 to 12.1 cm
Unit Size:
Pack of 1000
Pack of 500
Product Code. 30020313
Brand: Keystone Cleanroom Products 10TK09
Only null left
Add to basket
Add to basket
Autoclavable 1421B Tyvek™ Mini-Muffs prevent dust and other particulates from entering orifices of any size.
  • Conveniently covers flasks, beakers, containers, media bottles, valves, pipes and hoses
  • Prevents dust and other particulate from entering orifices of any size
  • Autoclavable - can be used during sterilization to reduce time consuming wrapping
  • Breathable, while providing a superior barrier post sterilization
  • Tyvek™ has the highest microbial barrier properties among all steam sterilization wrapping compatible materials
  • Manufactured and packaged in a Controlled Cleanroom Environment
  • Resists tearing and punctures
  • Eliminates Autoclave Tape Residue
  • Made from 1421B Tyvek Material
  • Latex-free rubber
  • Hydrophobic
  • Lint-Free
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Closure Type None
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
For Use With (Equipment) Pharmaceutical Equipment and Components
Material Tyvek
Sterility Non-sterile
Temperature Range (English) Up to 250°F
Temperature Range (Metric) Up to 125°C
Length (English) 4 to 5.5 in.
Length (Metric) 10.2 to 14 cm
Width (English) 4 to 5.5 in.
Width (Metric) 10.2 to 14 cm
Product Type Cover