Learn More
Cole-Parmer™ PVDF Union T-Connector Compression Fittings
Product Code. 11759529
Click to view available options
For Use With:
1/4 in. tubing O.D.
1/8 in. tubing O.D.
Port Size:
3/32 in. orifice, 1-1/8 x 3/4 in.
7/32 in. orifice, 1-15/32 x 1-1/32 in.
Width (Metric):
19 mm
26 mm
Unit Size:
Each
Product Code. 11759529
Brand: Cole-Parmer™ 0638245
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Compression fittings work well with rigid tubing for more permanent installation in high-pressure applications.
  • No leaks, flow restrictions or costly downtime
  • Fluid paths in the laboratory assembled without the use of tools
  • Provided with nut that compresses rigid tubing against fitting body
  • Sterilization by autoclave or ethylene oxide (ETO)
  • Offering excellent chemical resistance
Specifications
Autoclavable Autoclavable
Material PVDF
For Use With 1/8 in. tubing O.D.
Packaging Quantity 1 Each
Resistance Properties Excellent chemical resistance
Port Size 3/32 in. orifice, 1-1/8 x 3/4 in.
Sterilization Method Autoclave or Ethylene Oxide
Width (Metric) 19 mm
Product Type Compression Fitting