Learn More
Keystone Cleanroom Products Autoclavable Tyvek™ Stopper Tubing with Heat Seal Closure
Product Code. 17871711 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Dimensions (L x W):
18 in. x 500 ft.
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
17871711 18 in. x 500 ft. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17871711 Supplier Keystone Cleanroom Products Supplier No. 018SBT
Tyvek™ Stopper Tubing will not rip or puncture. Includes heat-seal closure and double-welded factory seals.
The medical Grade 1073B Tyvek™ material has the highest microbial barrier properties of any steam sterilization compatible materials. Tyvek Tubing is also compatible with VHP Decontamination cycles commonly used with Isolator technology. Will not absorb or adsorb water.
  • Manufactured and packaged in an ISO certified cleanroom
  • Double Welded Factory Seals
  • Autoclavable at up to 125°C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
  • Heat seals closed
  • Tubing provides sizing flexibility
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Closure Type Heat Seal
Color Clear, White
Disposable Single-use
For Use With (Application) Steam, EtO, Gamma or VHP Sterilization
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Quantity 500 Pk.
Dimensions (L x W) 18 in. x 500 ft.
Length (English) 500 ft.
Width (English) 18 in.
Product Type Sterilization Reel
Length (Metric) 152 m
Width (Metric) 45.7 cm