Learn More
Biochrom Automatic Sipper System
Click to view available options
Type:
Automatic Sipper System
Unit Size:
Each
Description
- Automatic aspiration
- User defined air gap and sip volume
- Waste or return option
- Local or PC-controlled
- Ability to calibrate sipped volume
- Choice of tubing kits
- Requires Tubing kit 80-7100-37 or 80-7100-38 and suitable flowcell 80-7100-40
Specifications
Specifications
| Type | Automatic Sipper System |
| For Use With (Equipment) | Ultrospec 7000/8000/9000 |