Learn More
Thermo Scientific™ Automation Tips for BioTek™ Pipetting Systems
Product Code. 16466949
Click to view available options
Volume (Metric):
50 μL
80 μL
100 μL
150 μL
200 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Unit Size:
Pack of 4800
Product Code. 16466949
Brand: Thermo Scientific™ 984262
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: BioTek Precision and Precision XS.
  • Available in non-sterile, sterile and with Thermo Scientific™ ART™ filter technology
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Format Rack
Tip Style Low Retention
For Use With (Equipment) BioTek Pipetting System
Product Line BioTek