Learn More
Thermo Scientific™ Autosampler Sample Tube and Standards Racks
Product Code. 17881145 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
iSC-65,CETAC, ESI
Includes:
60-Position Sample Tube Rack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Includes unitSize
17881145 iSC-65,CETAC, ESI 60-Position Sample Tube Rack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17881145 Supplier Thermo Scientific™ Supplier No. BRE0033171
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Customize your autosampler with a range of autosampler sample tube and standards racks to suit your requirements.
Match Your CETAC Sample Tube Rack to Your Laboratory’s Needs
  • 21-position sample tube rack
  • 24-position sample tube rack
  • 40-position sample tube rack
  • 60-position sample tube rack
  • 90-position sample tube rack
  • Extended Rack for CETAC ASX-520 Autosampler (for use with Thermo Scientific™ iCAP™ Q Series ICP-MS systems)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) iSC-65,CETAC, ESI
Type Autosampler Sample Tube Racks
Includes 60-Position Sample Tube Rack