Learn More
Hamilton™ Autosampler Syringes
Click to view available options
For Use With (Equipment):
AS200 Autosampler
Thermo-Finnigan / Fisons AS800/2000
Unit Size:
Each
Specifications
Specifications
| Material | Stainless Steel (Plunger) |
| Needle Gauge | 25s G |
| Diameter (Metric) Inner | 0.48 mm |
| Diameter (Metric) Outer | 6.6 mm |
| Product Type | Autosampler Syringe |
| Model | 701 |
| Needle Point Number | 2 |
| Quantity | 1/Ea. |
| Volume (Metric) | 10 μL |
| Needle Point Style | 2 |
| Show More |