Learn More
Trajan Scientific and Medical™ Autosampler Syringes with Cone Tipped Needle for Agilent GCs
Click to view available options
Volume (Metric):
1 μL
10 μL
Unit Size:
Each
Pack of 10
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Agilent instruments
Specifications
Specifications
| Needle Gauge | 23 G |
| Diameter (Metric) Inner Needle | 0.11 mm |
| Diameter (Metric) Outer Needle | 0.63 mm |
| Product Type | Removable Needle Agilent Syringe with GT Plunger |
| Volume (Metric) | 10 μL |
| Needle Point Style | Cone |
Product Content Correction
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion