Learn More
Trajan Scientific and Medical™ Autosampler Syringes with Cone Tipped Needle for Agilent GCs
Click to view available options
Volume (Metric):
1 μL
10 μL
Unit Size:
Each
Pack of 10
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Agilent instruments
Specifications
Specifications
| Needle Gauge | 23 G |
| Diameter (Metric) Inner Needle | 0.22 mm |
| Diameter (Metric) Outer Needle | 0.63 mm |
| Product Type | NanoVolume Agilent Syringe |
| Volume (Metric) | 1 μL |
| Needle Point Style | Cone |