Learn More
Trajan Scientific and Medical™ Autosampler Syringes with Fixed Needle for Thermo Scientific GCs
Product Code. 15238463
Click to view available options
Volume (Metric):
5 μL
10 μL
Needle Point Style:
Bevel
Cone
Product Type:
Fixed Needle
Removable Needle
Unit Size:
Each
Product Code. 15238463
Brand: Trajan Scientific and Medical™ 0029781
Gives better performance and superior quality. Trajan Scientific and Medical Autosampler Syringes with Fixed Needle for Thermo Scientific GCs has the knowledge and expertise to manufacture an SGE syringe.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Description CTC/Thermo (Classic Button) Syringe
Needle Gauge 26 G
Diameter (Metric) Outer Needle 0.47 mm
Product Type Fixed Needle
Volume (Metric) 10 μL
Needle Point Style Cone
For Use With (Equipment) Thermo Gas Chromatography