Learn More
Cytiva AutoSeq G-50 Columns
Shop All Cytiva Products
Click to view available options
Quantity:
1000 Columns
250 Columns
50 Columns
Unit Size:
Pack of 1000
Pack of 250
Pack of 50
Description
- Columns are prepacked with Sephadex G-50 DNA Grade F and pre-equilibrated in double-distilled water with 0.05% Kathon CG/ICP Biocide added as a preservative
- Columns that are ready to use enable sample application to collection of purified product in less than 4 min.
- Can be used to clean up reactions prepared with the BigDye Terminator v3.1 Cycle Sequencing Kit from Applied Biosystems
- Typical sample volume 12 to 25 μl
- Suitable for purification of DNA greater than 20 bases in length
- Optimized for automated fluorescent sequencing
Specifications
Specifications
| For Use With (Application) | Rapid removal of unincorporated Fluorescent dye-terminators from cycle sequencing reactions prior to automated sequencing |
| Product Type | Column |
| Quantity | 250 Columns |