missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Auxin response factor Monoclonal Antibody (14D2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA534937
This item is not returnable.
View return policy
Description
Positive Control: Tea leafbud.
Auxin response factors (ARFs) are transcription factors that bind to auxin response elements in promoters of early auxin response genes and play central roles in many auxin-mediated processes. As transcription factors, ARFs possess a modular structure and consist of three major domains: an amino-terminal DNA-binding domain (DBD); a middle domain, which is responsible for gene activation or repression; and a carboxy-terminal PB1(Phox and Bem1) domain, contained within a region that was previously called domain III/I. ARFs function as transcription activators or repressors, depending on the middle region of the ARF protein; Q-rich middle regions activate transcription, whereas P/S/T-rich middle regions confer repressor activity.
Specifications
| Auxin response factor | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Recombinant protein within tea Auxin response factor aa 401-586. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 14D2 | |
| Unconjugated | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| Plant | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction