missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Influenza A H5N1 Hemagglutinin Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41050C
This item is not returnable.
View return policy
Description
Avian Influenza A H5N1 Hemagglutinin Polyclonal antibody specifically detects Avian Influenza A H5N1 Hemagglutinin in Virus samples. It is validated for ELISA
Specifications
| Avian Influenza A H5N1 Hemagglutinin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| DyLight 650 | |
| Harvey rat sarcoma viral oncogene homolog | |
| Antibody was raised against a synthetic peptide corresponding to 14 amino acids in the middle of the avian influenza hemagglutinin protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within amino acids 320 - 370 of Avian Influenza Hemagglutinin. Amino Acid Squence: RNSPQRERRRKKRG | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction