missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Influenza A H5N1 Hemagglutinin Antibody, HRP, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-41066H
This item is not returnable.
View return policy
Description
Avian Influenza A H5N1 Hemagglutinin Polyclonal antibody specifically detects Avian Influenza A H5N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Avian Influenza A H5N1 Hemagglutinin | |
| Polyclonal | |
| Western Blot, ELISA | |
| Harvey rat sarcoma viral oncogene homolog | |
| Antibody was raised against a synthetic peptide corresponding to 10 amino acids near the center of the Hemagglutinin protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within amino acids 260 - 310 of Avian Influenza Hemagglutinin 4. Amino Acid Squence: REDIMKSELEYGN | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| Virus, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| HRP | |
| PBS | |
| Goat | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction