missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Influenza A H5N1 Neuraminidase Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41048IR
This item is not returnable.
View return policy
Description
Avian Influenza A H5N1 Neuraminidase Polyclonal antibody specifically detects Avian Influenza A H5N1 Neuraminidase in Virus samples. It is validated for ELISA
Specifications
| Avian Influenza A H5N1 Neuraminidase | |
| Polyclonal | |
| ELISA | |
| 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein | |
| Antibody was raised against a synthetic peptide corresponding to 16 amino acids in the middle of the avian influenza neuraminidase protein. Efforts were made to use relatively conserved regions as the antigen. The immunogen is located within amino acids 150 - 200 of Avian Influenza Neuraminidase. Amino Acid Squence: CPIGESPSPYNSRFES | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| Virus | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction