missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Influenza A H7N9 Neuraminidase Antibody - (A/Shanghai/02/2013) - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41280-0.025mg
This item is not returnable.
View return policy
Description
Avian Influenza A H7N9 Neuraminidase Polyclonal specifically detects Avian Influenza A H7N9 Neuraminidase in Virus samples. It is validated for ELISA.
Specifications
| Avian Influenza A H7N9 Neuraminidase | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| virus | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| ELISA | |
| 1 mg/mL | |
| ELISA 1 μg/mL | |
| EC=3.2.1.18, Neuraminidase | |
| Antibody was raised against a synthetic peptide corresponding to 17 amino acids near the amino terminus of the H7N9 [Influenza A virus (A/Shanghai/02/2013(H7N9))] Neuraminidase protein. The immunogen is located within amino acids 120 - 170 of Avian Influenza A H7N9 Neuraminidase. Amino Acid Squence: RGKHSNGTIHDRSQYRA | |
| 0.025 mg | |
| Primary | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction