missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Influenza A H7N9 Neuraminidase Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-41280CL1
This item is not returnable.
View return policy
Description
Avian Influenza A H7N9 Neuraminidase Polyclonal antibody specifically detects Avian Influenza A H7N9 Neuraminidase in Virus samples. It is validated for ELISA
Specifications
| Avian Influenza A H7N9 Neuraminidase | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA | |
| CoraFluor 1 | |
| EC=3.2.1.18, Neuraminidase | |
| Antibody was raised against a synthetic peptide corresponding to 17 amino acids near the amino terminus of the H7N9 [Influenza A virus (A/Shanghai/02/2013(H7N9))] Neuraminidase protein. The immunogen is located within amino acids 120 - 170 of Avian Influenza A H7N9 Neuraminidase. Amino Acid Squence: RGKHSNGTIHDRSQYRA | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction