Learn More
Avian Influenza NS1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-41067
 View more versions of this product
Product Code. 18117877
  • € 507.00 / 0.10mg
Description
Avian Influenza NS1 Polyclonal antibody specifically detects Avian Influenza NS1 in Virus samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Avian Influenza NS1
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
Antibody was raised against a synthetic peptide corresponding to 14 amino acids at the carboxy terminus of the Avian Influenza Nonstructural Protein 1 protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within the last 50 amino acids of Avian Influenza Nonstructural Protein 1. Amino Acid Squence: LPPNQKRKMARTIE
0.1 mg
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
ELISA
1 mg/mL
ELISA
Rabbit
Affinity Purified
RUO
Virus