missing translation for 'onlineSavingsMsg'
Learn More

Avian Influenza NS2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 168.00 - € 507.00

Specifications

Antigen Avian Influenza NS2
Concentration 1 mg/mL
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18624066
View Documents
Novus Biologicals
NBP2-41068-0.025mg
0.025 mg
€ 168.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18188307
View Documents
Novus Biologicals
NBP2-41068
0.1 mg
€ 507.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Avian Influenza NS2 Polyclonal antibody specifically detects Avian Influenza NS2 in Virus samples. It is validated for ELISA.
Specifications

Specifications

Avian Influenza NS2
ELISA
Unconjugated
RUO
PBS with 0.02% Sodium Azide
IgG
Affinity Purified
1 mg/mL
Polyclonal
Rabbit
Virus
Antibody was raised against a synthetic peptide corresponding to 14 amino acids near the middle of the Avian Influenza Nonstructural Protein 2 protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within amino acids 40 - 90 of Avian Influenza Nonstructural Protein 2. Amino Acid Squence: RNGKWREQLSQKFE
Primary
Videos
SDS
Documents

Documents

Product Certifications
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato