missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Influenza NS2 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41068V
This item is not returnable.
View return policy
Description
Avian Influenza NS2 Polyclonal antibody specifically detects Avian Influenza NS2 in Virus samples. It is validated for ELISA
Specifications
| Avian Influenza NS2 | |
| Polyclonal | |
| ELISA | |
| FIGLER6, SALM3 | |
| Antibody was raised against a synthetic peptide corresponding to 14 amino acids near the middle of the Avian Influenza Nonstructural Protein 2 protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within amino acids 40 - 90 of Avian Influenza Nonstructural Protein 2. Amino Acid Squence: RNGKWREQLSQKFE | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| Virus, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction