Learn More
Avian Influenza NS2 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-41068H
Product Code. 18663976
  • € 582.00 / 0.10mL
Description
Avian Influenza NS2 Polyclonal antibody specifically detects Avian Influenza NS2 in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Avian Influenza NS2
Polyclonal
ELISA
Rabbit
Peptide affinity purified
RUO
Primary
Store at 4C in the dark.
IgG
ELISA
HRP
PBS
Antibody was raised against a synthetic peptide corresponding to 14 amino acids near the middle of the Avian Influenza Nonstructural Protein 2 protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within amino acids 40 - 90 of Avian Influenza Nonstructural Protein 2. Amino Acid Squence: RNGKWREQLSQKFE
0.1 mL
Infections (Virus Bacteria and Parasites)
Virus