Learn More
KCTD14 Antibody (OTI2E9), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-72171B
Product Code. 18159022
  • € 751.00 / 0.10mL
Description
KCTD14 Monoclonal specifically detects KCTD14 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KCTD14
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
KCTD14
Full length recombinant protein of human KCTD14 (NP_076419) produced in HEK293T cells.
0.1 mL
65987.0
Store at 4°C in the dark.
Western Blot
OTI2E9
Western Blot
BTB/POZ domain-containing protein KCTD14, MGC2376, potassium channel tetramerisation domain containing 14
Mouse
Affinity Purified
Primary
Human
IgG2a