missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AVPI1 Monoclonal Antibody (OTI6C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802991S
This item is not returnable.
View return policy
Description
AVPI1 Monoclonal antibody specifically detects AVPI1 in Human samples. It is validated for Western BlotSpecifications
| AVPI1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P11362 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2260 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI6C5 | |
| Unconjugated | |
| FGFR1 | |
| arginine vasopressin induced 1; arginine vasopressin-induced 1; arginine vasopressin-induced protein 1; AVPI1; AVP-induced protein 1; NPD013; PP5395; RP11-548K23.7; vasopressin-induced transcript; VIP32; VIT32 | |
| Human recombinant protein fragment corresponding to amino acids 1-376 of human FGFR1 produced in SF9 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction