missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axin-1 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18242AF405
This item is not returnable.
View return policy
Description
Axin-1 Polyclonal antibody specifically detects Axin-1 in Mouse samples. It is validated for Western Blot
Specifications
| Axin-1 | |
| Polyclonal | |
| Western Blot | |
| axin 1, axin-1, AXINaxis inhibitor 1, Axis inhibition protein 1, fused, mouse, homolog of, hAxin, MGC52315 | |
| Synthetic peptide of Human Axin-1 (200-300 aa), conjugated to Keyhole Limpet Haemocyanin (KLH). | |
| 0.1 mL | |
| Apoptosis, Wnt Signaling Pathway | |
| 8312 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction