Learn More
Tocris Bioscience™ Axitinib
GREENER_CHOICE
Product Code. 18035832
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18035832 10 mg 10mg
18015112 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18035832 Supplier Tocris Bioscience™ Supplier No. 4350/10
Potent VEGFR-1, -2 and -3 inhibitor
Axitinib is a potent inhibitor of VEGFR-2, -3, and -1 (IC50 values are 0.2, 0.1-0.3, and 1.2 nM respectively). Axitinib exhibits minimal activity against a panel of ∼100 protein kinases. Axitinib inhibits angiogenesis and vascular permeability. Also a high affinity BCR-ABL1 (T315I) inhibitor (Ki = 149 pM for autophosphorylated ABL1 (T315I)). Axitinib blocks proliferation of Ba/F3 cells expressing BCR-ABL1 (T315I). Axitinib (10 mg/kg) decreases T-cell exhaustion and tumor and T-cell hypoxia and improves survival in mice in vivo. Orally available.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 319460-85-0
Molecular Formula C22H18N4OS
Specifications
Quantity 10 mg
Formula Weight 386.47
Purity >99%
Molecular Weight (g/mol) 386.473
Chemical Name or Material Axitinib