missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axl Antibody (108724R) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB154RJF585
This item is not returnable.
View return policy
Description
Axl Monoclonal antibody specifically detects Axl in Human samples. It is validated for Flow Cytometry,ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair),CyTOF
Specifications
| Axl | |
| Monoclonal | |
| Janelia Fluor 585 | |
| AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, tyrosine-protein kinase receptor UFO, UFOoncogene AXL | |
| Mouse myeloma cell line NS0-derived recombinant human Axl, Met1-Pro440, Accession # AAA61243 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF | |
| 108724R | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cell Cycle and Replication, Phospho Specific | |
| 558 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction