missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axl Antibody (175128) [DyLight 550], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB8541L
This item is not returnable.
View return policy
Description
Axl Monoclonal antibody specifically detects Axl in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Axl | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 175128 | |
| Flow Cytometry, CyTOF-ready | |
| AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, tyrosine-protein kinase receptor UFO, UFOoncogene AXL | |
| Mouse myeloma cell line NS0-derived recombinant mouse Axl, Ala19-Trp445, Accession # Q00993 | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 558 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction