missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axl Antibody (MM0098-2N33), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12044PE
This item is not returnable.
View return policy
Description
Axl Monoclonal specifically detects Axl in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Cell Surface).
Specifications
| Axl | |
| Monoclonal | |
| PE | |
| AAA61243 | |
| AXL | |
| Human recombinant Axl EC domain | |
| 0.1 mL | |
| Cell Cycle and Replication, Phospho Specific | |
| 558 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| MM0098-2N33 | |
| Flow Cytometry | |
| AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, tyrosine-protein kinase receptor UFO, UFOoncogene AXL | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction