Learn More
Axygen™ 96-well Low Profile Bar Coded PCR Microplates
Shop All Axygen Scientific Products
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
Description
- Low profile; Optimized for fast PCR
- Certified RNase-/DNase-free
- Nonpyrogenic, nonsterile
- Bar coded for traceability and identification
- Sealing options: Axymats, heat sealing, adhesive films and foils, flat and domed cap strips
Specifications
Specifications
| Barcode | Includes Barcode |
| Certifications/Compliance | RNase-/DNase-free |
| Format | Standard |
| Array | 8 x 12 |
| Color | Clear |
| For Use With (Equipment) | ABI thermal cyclers, ABI Fast PCR 7500, 7900 9800, Veriti and StepOne |
| Material | Polypropylene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| For Use With (Application) | PCR thermal transfer, RT-PCR applications |