Learn More
Axygen™ Electrical Leads for 7 cm Gel Box
Product Code. 15573097 Shop All Axygen Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Gel Box, 7 cm
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15573097 Gel Box, 7 cm 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15573097 Supplier Axygen™ Supplier No. HGBEL
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Accessory for use with 7cm horizontal gel box
1 Pair
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Gel Box, 7 cm
Type Electrical Leads (1 Pair)