Learn More
Axygen™ Gel Casting Trays for 7 cm Gel Box
Product Code. 15583167 Shop All Axygen Scientific Products
Change view
Click to view available options
Dimensions (L x W):
7 x 10 cm (2.7 x 3.9 in.)
7 x 7 cm (2.7 x 2.7 in.)
Unit Size:
1 unit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15583167 7 x 7 cm (2.7 x 2.7 in.) 1 unit
15523167 7 x 10 cm (2.7 x 3.9 in.) 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15583167 Supplier Axygen™ Supplier No. HGB77GT
Estimated Shipment: 02-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Accessory for use with 7cm horizontal gel box
  • UV transparent
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Gel Box, 7 cm
Dimensions (L x W) 7 x 7 cm (2.7 x 2.7 in.)