Learn More
Axygen™ Polypropylene Microtubes with Blue Screw Cap
Shop All Axygen Scientific Products
Click to view available options
Capacity (Metric):
0.5 mL
Unit Size:
Pack of 500
Specifications
Specifications
| Cap Style | Screw Cap |
| Closure Color | Blue |
| Capacity (Metric) | 0.5 mL |
| Material | Polypropylene |
| Includes | Screw Cap, O-ring |
| No. per Pack | 4000Pack |
| Packaging | 4000Pack |
| Sterility | Sterile |
| Graduated | No |
| Closure Type | Screw Cap |