Learn More
Axygen™ Aerosol Barrier Micropipette Tips
Product Code. 12581348 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
0.5 to 50 μL
1 to 150 μL
100 to 1000 μL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1000
Pack of 10000
Pack of 4800
Pack of 960
Product Code. 12581348
Brand: Axygen™ TF150
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Axygen™ Aerosol Barrier Micropipette Tips allow free passage of air while blocking aerosol contamination.
  • Technologically advanced filter material
  • Filter membrane contains no leachable additives
  • Available with Maxymum Recovery™ surface for improved accuracy and maximum sample recovery
  • Available in bulk, or racked (sterile)
  • Available in a variety of material contact tip options
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 100 to 1000 μL
Filtered Filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase Free
Format Bulk
Tip Style Universal
Packaging Bulk = 1000 tips/pack
Product Type Aerosol Barrier Micropipette Tips
For Use With (Equipment) Universal Fit
Material Polypropylene