Learn More
Axygen™ Power Supply Adapters for 7cm Gel Box
Product Code. 15523177 Shop All Axygen Scientific Products
Change view
Click to view available options
Type:
4 mm to 2 mm Power Supply Adapter
4 mm to 4 mm Power Supply Adapter
Unit Size:
1 unit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15523177 4 mm to 2 mm Power Supply Adapter 1 unit
15513177 4 mm to 4 mm Power Supply Adapter 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15523177 Supplier Axygen™ Supplier No. HGB7AD2
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Accessory for use with 7cm horizontal gel box
Power supply adapters
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Gel Box, 7 cm
Type 4 mm to 2 mm Power Supply Adapter