Learn More
Axygen™ Replacement Electrodes for 7cm Gel Box
Product Code. 15553177 Shop All Axygen Scientific Products
Change view
Click to view available options
Type:
Replacement Negative Electrode
Replacement Positive Electrode
Unit Size:
1 unit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15553177 Replacement Negative Electrode 1 unit
15543177 Replacement Positive Electrode 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15553177 Supplier Axygen™ Supplier No. HGB7PE
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Accessory for use with 7cm horizontal gel box
Replacement electrode
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Gel Box, 7 cm
Type Replacement Negative Electrode