Learn More
Axygen™ Screw Cap Tubes without Caps: Self-Standing with Conical Bottoms
Shop All Axygen Scientific Products
Click to view available options
Color:
Clear
Sterility:
Sterile
Capacity (Metric):
1.5 mL
2 mL
Unit Size:
Pack of 4000
Description
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping
Specifications
Specifications
| Certifications/Compliance | RNase, DNase, pyrogen-free |
| Capacity (Metric) | 1.5 mL |
| Color | Clear |
| Material | Polypropylene |
| Packaging | Bag |
| Sterility | Sterile |
| Closure Type | Not Included |
| Bottom Shape | Conical |
| Max. RCF | 20,000 x g |