Learn More
Axygen™ Automation Micropipette Tips
Shop All Axygen Scientific Products
Click to view available options
Sterility:
Non-sterile
Sterile
Tip Style:
Automation, Maxymum Recovery, Round
Automation, Round
Unit Size:
Pack of 4800
Description
- For Tecan™/Freedom EVO&trade, Te-Mo™, Aquarius, Zymark™/Caliper, RapidPlate™, and SciClone™ Systems
- For 96-well, Non-ANSI and ANSI/SLAS (384) format, number 5 polypropylene racks
- Available with or without aerosol-resistant filters
- Available sterile/non-sterile
- Available in standard or Maxymum Recovery™ style
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Specifications
Specifications
| Volume (Metric) | 200 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Clear |
| Rack Color | Blue |
| Autoclavable | Autoclavable |
| Clean Claims | RNase-free, DNase-free, Nonpyrogenic |
| Format | Rack |
| Tip Style | Automation, Round |
| Packaging | 96 tips/rack, 10 racks/pack, 5 packs/case |
| Show More |