Learn More
Azamethiphos-d6, TRC
Specifications
Specifications
| Chemical Name or Material | Azamethiphos-d6 |
| CAS | 1189894-02-7 |
| Molecular Formula | C9 D6 H4 Cl N2 O5 P S |
| Quantity | 10 mg |
| Synonym | 3-[Bis(trideuteriomethoxy)phosphorylsulfanylmethyl]-6-chloro-[1,3]oxazolo[4,5-b]pyridin-2-one |
| InChI Formula | InChI=1S/C9H10ClN2O5PS/c1-15-18(14,16-2)19-5-12-8-7(17-9(12)13)3-6(10)4-11-8/h3-4H,5H2,1-2H3/i1D3,2D3 |
| SMILES | [2H]C([2H])([2H])OP(=O)(OC([2H])([2H])[2H])SCN1C(=O)Oc2cc(Cl)cnc12 |
| IUPAC Name | 3-[bis(trideuteriomethoxy)phosphorylsulfanylmethyl]-6-chloro-[1,3]oxazolo[4,5-b]pyridin-2-one |
| Molecular Weight (g/mol) | 330.71 |
| Formula Weight | 330.0113 |