Learn More
AZD4547, MedChemExpress
Product Code. p-200096425
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Product Code. 16495648
Brand: Medchem Express HY13330
MedChemExpress AZD4547 is a potent inhibitor of the FGFR family with IC50s of 0.2 nM, 2.5 nM, 1.8 nM, and 165 nM for FGFR1, FGFR2, FGFR3, and FGFR4, respectively.
AZD4547 is a potent inhibitor of the FGFR family with IC50s of 0.2 nM, 2.5 nM, 1.8 nM, and 165 nM for FGFR1, FGFR2, FGFR3, and FGFR4, respectively.
Specifications
Chemical Name or Material AZD4547
CAS 1035270-39-3
Color White
Purity Grade Notes Research
Molecular Formula C26H33N5O3
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (269.65 mM; Need ultrasonic)
SMILES C[C@H](C1)N[C@@H](C)CN1C2=CC=C(C(NC3=NNC(CCC4=CC(OC)=CC(OC)=C4)=C3)=O)C=C2
Molecular Weight (g/mol) 463.57
Formula Weight 463.57
Percent Purity 98.87%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only