Learn More
AZD5153 6-Hydroxy-2-naphthoic acid, MedChemExpress
Product Code. 16479666
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16479666
Brand: Medchem Express HY100653A
MedChemExpress AZD5153 6-Hydroxy-2-naphthoic acid is the 6-Hydroxy-2-naphthoic acid of AZD5153. AZD5153 is a potent, selective, and orally available BET/BRD4 bromodomain inhibitor; disrupts BRD4 with an IC50 of 1.7 nM.
AZD5153 6-Hydroxy-2-naphthoic acid is the 6-Hydroxy-2-naphthoic acid of AZD5153. AZD5153 is a potent, selective, and orally available BET/BRD4 bromodomain inhibitor
Specifications
Chemical Name or Material AZD5153 6-Hydroxy-2-naphthoic acid
CAS 1869912-40-2
Color White
Purity Grade Notes Research
Molecular Formula C36H41N7O6
Quantity 50 mg
Synonym AZD-5153 HNT salt
Solubility Information DMSO : ≥ 34 mg/mL (50.92 mM)
SMILES O=C(C1=CC=C2C=C(O)C=CC2=C1)O.O=C3N(C)CCN(CCOC4=CC=C(C5CCN(C6=NN7C(C=C6)=NN=C7OC)CC5)C=C4)[C@@H]3C
Molecular Weight (g/mol) 667.75
Formula Weight 667.75
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only