missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZI2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-97191CL1
This item is not returnable.
View return policy
Description
AZI2 Polyclonal antibody specifically detects AZI2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| AZI2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 64343 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| 5-azacytidine induced 2, AZ25-azacytidine-induced protein 2, FLJ21939, NAP15-azacytidine induced gene 2, NF-kappa-B-activating kinase-associated protein 1, TILPNak-associated protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human AZI2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction