missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AZI2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16692965 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16692965 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16692965 Supplier Invitrogen™ Supplier No. PA5106532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AZI2.

AZI2, or NAP1, contributes to the activation of NFKB (Fujita et al., 2003 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen AZI2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AZI2
Gene Accession No. Q4KMA0, Q9H6S1, Q9QYP6
Gene Alias 5-azacytidine induced 2; 5-azacytidine induced gene 2; 5-azacytidine-induced protein 2; AA410145; Az2; AZI2; Nak-associated protein 1; NAP1; NF-kappa-B-activating kinase-associated protein 1; TBKBP2; Tbkp2; TILP
Gene Symbols AZI2
Host Species Rabbit
Immunogen A synthesized peptide derived from human AZI2(Accession Q9H6S1), corresponding to amino acid residues M1-V42.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27215, 316051, 64343
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.