missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZIN1 Monoclonal Antibody (OTI5C7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810908
This item is not returnable.
View return policy
Description
Antizyme inhibitor (AZIN1) protein that positively regulates ornithine decarboxylase (ODC) activity and polyamine uptake. AZI is an enzymatically inactive ODC homolog that counteracts the negative effect of ODC antizymes (AZs) OAZ1, OAZ2 and OAZ3 on ODC activity by competing with ODC for antizyme-binding. Inhibits antizyme-dependent ODC degradation and releases ODC monomers from their inactive complex with antizymes, leading to formation of the catalytically active ODC homodimer and restoring polyamine production.Specifications
| AZIN1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14977, Q63764 | |
| AZIN1 | |
| Human recombit protein fragment corresponding to amino acids 260-448 of human AZIN1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5C7 | |
| Unconjugated | |
| AZIN1 | |
| AZI, AZI1, AZIA1, OAZI, OAZIN, ODC1L | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51582, 58961 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction