missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ LDPE Colored Narrow Neck Wash Bottles

Catalog Number 10257874
Click to view available options
Color:
Green
Red
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Use these durable plastic bottles for a wide range of liquid storage, media preparation and sampling applications. Azlon™ LDPE Colored Narrow Neck Wash Bottles are fitted with matching polypropylene closures. Ideal for color-coding purposes.

  • Colored wash bottles with matching closures
  • Fitted with polypropylene closures
  • Choice of four colors
Closure Material Polypropylene
Height (Metric) 160 mm
Capacity (Metric) 500 mL
Color Green
For Use With (Application) For color coding purposes
Includes Polypropylene matching closures
Material LDPE
Quantity 5/PK
Temperature Range (Metric) –50 to 80°C
Closure Included Yes
Autoclavable Not Autoclavable
Mouth Narrow
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'