Learn More
Azlon™ Magnetic Stir Bar
Click to view available options
Length (Metric):
15 mm
Unit Size:
Pack of 2
Description
- For efficient stirring
- Cylindrical shaped
Specifications
Specifications
| Color | PTFE |
| Diameter (Metric) | 6 mm |
| Length (Metric) | 15 mm |
| Quantity | 2 |
| Shape | Cylinder |