Learn More
Azlon™ Polypropylene Class B Cylinder Set
Click to view available options
Unit Size:
Each
Description
- With molded grads
Specifications
Specifications
| Material | Polypropylene |
| Includes | 10, 25, 50, 100, 250 and 500 mL Cylinders |
| Quantity | 1/Pk. |
| Graduations | Yes |
| Type | Ungraduated Cylinder Set |