Learn More
Azlon™ Polypropylene Powder Funnels
Product Code. 12040783
Click to view available options
Diameter (Metric)Top:
65mm
80mm
100mm
120mm
150mm
290mm
420mm
Diameter (Metric) Stem:
18mm
22mm
26mm
30mm
35mm
36mm
48mm
Height (Metric):
69 mm
74 mm
90 mm
106 mm
135 mm
208 mm
270 mm
Unit Size:
Each
Pack of 5
Product Code. 12040783
Brand: Azlon FWC214
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Constructed of durable, lightweight Polypropylene. Azlon™ Polypropylene Powder Funnel features a parallel stem that minimises 'bridging' of powder to improve flow. The two largest sizes have vertical-sided tops to prevent spillage.
  • Lightweight polypropylene powder funnel
  • Unbreakable in normal use
  • Parallel stem minimises 'bridging' of powder to improve flow
  • The two largest sizes have vertical-sided tops
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 420mm
Diameter (Metric) Stem 48mm
Material Polypropylene
Height (Metric) 270 mm