missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AZM 475271
GREENER_CHOICE

Product Code. 18054792
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18054792

Brand: Tocris Bioscience™ 3963/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Src tyrosine kinase inhibitor

Src tyrosine kinase inhibitor. Reduces tumor size, vascularity and metastasis, and increases apoptosis in human pancreatic cells grown in nude mice. Also exhibits antiangiogenic activity in vitro and in vivo. Sensitizes tumor cells to the cytotoxic effects of gemcitabine (Cat. No. 3259).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 476159-98-5
Molecular Formula C23H27ClN4O3

Specifications

Quantity 50 mg
Formula Weight 442.94
Purity >99%
Molecular Weight (g/mol) 442.944
Chemical Name or Material AZM 475271
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.