Learn More
Azocarmine G, TRC
Specifications
Specifications
| Chemical Name or Material | Azocarmine G |
| CAS | 25641-18-3 |
| Molecular Formula | C28 H18 N3 O6 S2 . Na |
| Quantity | 250 mg |
| Synonym | Benzo[a]phenazinium, 7-phenylsulfo-5-[(4-sulfophenyl)amino]-, hydroxide, inner salt, monosodium salt, Benzo[a]phenazinium, 7-phenylsulfo-5-[(4-sulfophenyl)amino]-, inner salt, monosodium salt (9CI), C.I. Acid Red 101 (7CI), C.I. Acid Red 101, monosodium s |
| InChI Formula | InChI=1S/C28H19N3O6S2.Na/c32-38(33,34)20-12-10-18(11-13-20)29-25-17-27-28(22-15-14-21(16-23(22)25)39(35,36)37)30-24-8-4-5-9-26(24)31(27)19-6-2-1-3-7-19;/h1-17H,(H2,32,33,34,35,36,37);/q;+1/p-1 |
| SMILES | [Na+].[O-]S(=O)(=O)c1ccc(Nc2cc3c(nc4ccccc4[n+]3c5ccccc5)c6ccc(cc26)S(=O)(=O)[O-])cc1 |
| IUPAC Name | sodium;7-phenyl-5-(4-sulfonatoanilino)benzo[a]phenazin-7-ium-3-sulfonate |
| Molecular Weight (g/mol) | 579.579 |
| Formula Weight | 579.053 |