Learn More
Azure C (Technical Grade), TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Azure C (Technical Grade) |
| CAS | 531-57-7 |
| Molecular Formula | C13H14N3SCl |
| Quantity | 5 g |
| Synonym | Azure C (6CI), Phenothiazin-5-ium, 3-amino-7-(methylamino)-, chloride (8CI,9CI), 3-Amino-7-methylaminophenazathionium chloride, Monomethylthionine, Monomethylthionine chloride |
| InChI Formula | InChI=1S/C13H12N3S.ClH/c1-15-9-3-5-11-13(7-9)17-12-6-8(14)2-4-10(12)16-11;/h2-7,15H,14H2,1H3;1H/q+1;/p-1 |
| SMILES | [Cl-].CNc1ccc2nc3ccc(N)cc3[s+]c2c1 |
| IUPAC Name | 7-N-methylphenothiazin-5-ium-3,7-diamine;chloride |
| Molecular Weight (g/mol) | 277.77 |
| Formula Weight | 277.044 |